Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL1 | NM_001128150.1 | NP_001121622.1 | ||
| NM_016467.4 | NP_057551.1 | |||
| XM_005246970.2 | XP_005247027.1 | |||
| XM_011512199.2 | XP_011510501.1 | |||
| XM_011512200.2 | 1 | 659 | XP_011510502.1 | |
| XM_017005347.1 | XP_016860836.1 | |||
| XM_017005348.1 | XP_016860837.1 | |||
| AF151036.1 | AAF36122.1 | |||
| AF395704.1 | AAM43503.1 | |||
| AK074756.1 | BAC11184.1 | |||
| AK313533.1 | ||||
| BC005200.1 | AAH05200.1 | |||
| DB122266.1 |
More Information
Set Membership: |
|

Genomic Map