Assay Details
Target Gene Details
Entrez Gene ID: | 151242 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 1C |
Gene Aliases: |
IPP5 |
Location: |
Chr.2:181954241-182131382 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R1C | NM_001080545.2 | NP_001074014.1 | ||
| NM_001261424.1 | NP_001248353.1 | |||
| NM_001261425.1 | NP_001248354.1 | |||
| NR_048566.1 | ||||
| NR_048567.1 | ||||
| AF494535.1 | AAM18054.1 | |||
| AK129898.1 | ||||
| AK311571.1 | ||||
| BC017943.1 | AAH17943.1 | |||
| BX647581.1 | CAI46089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834479 | Chr.2:181873756 - 182040717 on Build GRCh38 | Gain |
|
| esv3893515 | Chr.2:181940873 - 182006405 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map