Assay Details
Target Gene Details
Entrez Gene ID: | 100271715 |
Gene Name: | Rho guanine nucleotide exchange factor 33 |
Gene Aliases: |
- |
Location: |
Chr.2:38919363-38975449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF33 | NM_001145451.2 | NP_001138923.2 | ||
| DV080278.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114377135] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map