Assay Details
Target Gene Details
Entrez Gene ID: | 100996579 |
Gene Name: | uncharacterized LOC100996579 |
Gene Aliases: |
- |
Location: |
Chr.2:161244739-161249050 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100996579 | NR_110163.1 | |||
| NR_110164.1 | ||||
| AK027541.1 | ||||
| BG697582.1 |
Target Gene Details
Entrez Gene ID: | 101929512 |
Gene Name: | uncharacterized LOC101929512 |
Gene Aliases: |
- |
Location: |
Chr.2:161223258-161254643 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929512 | NR_110593.1 | |||
| BX107690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7140n54 | Chr.2:161155624 - 161270523 on Build GRCh38 | Loss |
|
| nsv834437 | Chr.2:161075774 - 161268203 on Build GRCh38 | Gain |
|
| nsv955277 | Chr.2:161245790 - 161282589 on Build GRCh38 | Duplication |
|
| nsv459934 | Chr.2:161203306 - 161389321 on Build GRCh38 | Loss |
|

Genomic Map