Assay Details
Target Gene Details
Entrez Gene ID: | 375190 |
Gene Name: | family with sequence similarity 228 member B |
Gene Aliases: |
- |
Location: |
Chr.2:24076874-24169638 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM228B | NM_001291328.1 | NP_001278257.1 | ||
| NR_111929.1 | ||||
| BM722814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv2641 | Chr.2:24081872 - 24103374 on Build GRCh38 | Deletion |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map