Assay Details
Target Gene Details
Entrez Gene ID: | 9577 |
Gene Name: | brain and reproductive organ-expressed (TNFRSF1A modulator) |
Gene Aliases: |
BRCC4, BRCC45 |
Location: |
Chr.2:27890615-28338901 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRE | NM_001261840.1 | NP_001248769.1 | ||
| NM_004899.4 | NP_004890.2 | |||
| NM_199191.2 | NP_954661.1 | |||
| NM_199192.2 | NP_954662.1 | |||
| NM_199193.2 | NP_954663.1 | |||
| NM_199194.2 | NP_954664.1 | |||
| AF015767.1 | AAB69387.1 | |||
| AF420602.1 | AAL17814.1 | |||
| AF420603.1 | AAL17816.1 | |||
| AF420604.1 | AAL17817.1 | |||
| AF420605.1 | AAL17818.1 | |||
| AK000097.1 | BAA90943.1 | |||
| AK291086.1 | ||||
| BG422104.1 | ||||
| BG422293.1 | ||||
| L38616.1 | AAA64231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759034 | Chr.2:27838469 - 28001763 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map