Assay Details
Target Gene Details
Entrez Gene ID: | 100128590 |
Gene Name: | SLC8A1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:39917634-40255209 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8A1-AS1 | NR_038441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590441 | Chr.2:39864446 - 40005186 on Build GRCh38 | Loss |
|
| nsv1006550 | Chr.2:39937629 - 39982796 on Build GRCh38 | Gain |
|
| dgv449n27 | Chr.2:39924742 - 39981525 on Build GRCh38 | Loss |
|
| esv3575235 | Chr.2:39918775 - 40018324 on Build GRCh38 | Gain |
|
| dgv6794n54 | Chr.2:39927704 - 40014728 on Build GRCh38 | Gain |
|
| dgv3791n100 | Chr.2:39914636 - 40018324 on Build GRCh38 | Gain |
|
| esv3891871 | Chr.2:39843084 - 40072324 on Build GRCh38 | Loss |
|
| nsv998885 | Chr.2:39896764 - 39955711 on Build GRCh38 | Loss |
|
| nsv1003896 | Chr.2:39866941 - 40004445 on Build GRCh38 | Loss |
|

Genomic Map