Assay Details
Target Gene Details
Entrez Gene ID: | 841 |
Gene Name: | caspase 8 |
Gene Aliases: |
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5 |
Location: |
Chr.2:201233443-201287711 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP8 | NM_001080124.1 | NP_001073593.1 | ||
| NM_001228.4 | NP_001219.2 | |||
| NM_033355.3 | NP_203519.1 | |||
| NM_033358.3 | NP_203522.1 | |||
| XM_005246887.1 | XP_005246944.1 | |||
| XM_005246888.1 | XP_005246945.1 | |||
| XM_005246894.3 | XP_005246951.1 | |||
| XM_006712789.1 | XP_006712852.1 | |||
| XM_006712793.2 | XP_006712856.1 | |||
| AF422927.1 | AAL87630.1 | |||
| AF422928.1 | AAL87631.1 | |||
| AK310630.1 | ||||
| BG498355.1 | ||||
| CB990598.1 | ||||
| DA420056.1 | ||||
| U60520.1 | AAC50645.1 | |||
| X98176.1 | CAA66857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954819 | Chr.2:201230778 - 201242577 on Build GRCh38 | Duplication |
|
| esv3893532 | Chr.2:201232852 - 201351096 on Build GRCh38 | Loss |
|
| nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76068885] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map