Assay Details
Target Gene Details
Entrez Gene ID: | 55515 |
Gene Name: | acid sensing ion channel subunit family member 4 |
Gene Aliases: |
ACCN4, BNAC4 |
Location: |
Chr.2:219514170-219538772 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC4 | NM_182847.2 | NP_878267.2 | ||
| XM_005246671.2 | XP_005246728.2 | |||
| XM_017004439.1 | XP_016859928.1 | |||
| AK308804.1 | ||||
| AK309373.1 | ||||
| BC010439.1 | AAH10439.1 | |||
| BC031812.1 | AAH31812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834545 | Chr.2:219500798 - 219589142 on Build GRCh38 | Loss |
|
| nsv584502 | Chr.2:219486657 - 219550995 on Build GRCh38 | Gain |
|
| nsv520958 | Chr.2:219404753 - 219565481 on Build GRCh38 | Loss |
|
| nsv508887 | Chr.2:219507073 - 219610784 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map