Assay Details
Target Gene Details
Entrez Gene ID: | 100271715 |
Gene Name: | Rho guanine nucleotide exchange factor 33 |
Gene Aliases: |
- |
Location: |
Chr.2:38919363-38975449 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF33 | NM_001145451.2 | 4 | 401 | NP_001138923.2 |
| DV080125.1 | 2 | 116 | ||
| DV080126.1 | 3 | 255 |
More Information
Set Membership: |
|

Genomic Map