Assay Details
Target Gene Details
Entrez Gene ID: | 100379249 |
Gene Name: | BOK antisense RNA 1 |
Gene Aliases: |
BOK-AS, BOKAS, NAToB, NCRNA00151 |
Location: |
Chr.2:241544384-241559143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOK-AS1 | NR_033346.1 | |||
| FJ185168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759120 | Chr.2:241480540 - 241562243 on Build GRCh38 | Loss |
|
| esv3344058 | Chr.2:241545364 - 241547362 on Build GRCh38 | Duplication |
|
| nsv834588 | Chr.2:241498269 - 241613859 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150194885] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map