Assay Details
Target Gene Details
Entrez Gene ID: | 9392 |
Gene Name: | transforming growth factor beta receptor associated protein 1 |
Gene Aliases: |
TRAP-1, TRAP1, VPS3 |
Location: |
Chr.2:105249404-105329714 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFBRAP1 | NM_001142621.2 | 12 | 5523 | NP_001136093.1 |
| NM_004257.5 | 12 | 5526 | NP_004248.2 | |
| XM_011512193.2 | XP_011510495.1 | |||
| XM_017005339.1 | 11 | 5019 | XP_016860828.1 | |
| BM672953.1 | 1 | 151 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012462 | Chr.2:105183092 - 105294801 on Build GRCh38 | Gain |
|
| nsv582577 | Chr.2:105167153 - 105302140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map