Assay Details
Target Gene Details
Entrez Gene ID: | 57624 |
Gene Name: | neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2 |
Gene Aliases: |
KIAA1486 |
Location: |
Chr.2:225399710-225703653 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NYAP2 | NM_020864.1 | NP_065915.1 | ||
| XM_005246708.3 | XP_005246765.1 | |||
| XM_011511524.1 | XP_011509826.1 | |||
| XM_017004564.1 | XP_016860053.1 | |||
| AB040919.1 | ||||
| AK055226.1 | BAB70880.1 | |||
| BC131727.1 | ||||
| DA230522.1 |
More Information
Set Membership: |
|

Genomic Map