Assay Details
Target Gene Details
Entrez Gene ID: | 100271715 |
Gene Name: | Rho guanine nucleotide exchange factor 33 |
Gene Aliases: |
- |
Location: |
Chr.2:38919363-38975449 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF33 | NM_001145451.2 | NP_001138923.2 | ||
| DV080660.1 |
Target Gene Details
Entrez Gene ID: | 375196 |
Gene Name: | uncharacterized LOC375196 |
Gene Aliases: |
- |
Location: |
Chr.2:38959287-38960344 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC375196 | NR_028386.1 | 1 | 725 | |
| BC030087.2 | 1 | 726 | AAH30087.2 |
More Information
Set Membership: |
|

Genomic Map