Assay Details
Target Gene Details
Entrez Gene ID: | 6936 |
Gene Name: | GC-rich sequence DNA-binding factor 2 |
Gene Aliases: |
C2orf3, DNABF, GCF, TCF9 |
Location: |
Chr.2:75662655-75710986 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCFC2 | NM_001201334.1 | NP_001188263.1 | ||
| NM_003203.4 | NP_003194.3 | |||
| XM_005264520.3 | XP_005264577.1 | |||
| XM_011533074.2 | XP_011531376.1 | |||
| AB026911.1 | BAA85386.1 | |||
| AK097564.1 | ||||
| BC064559.1 | AAH64559.1 | |||
| EF158468.1 | ||||
| EF158469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv706e214 | Chr.2:75581910 - 75690050 on Build GRCh38 | Gain |
|
| dgv20n14 | Chr.2:75596036 - 75687909 on Build GRCh38 | Gain |
|
| esv2759063 | Chr.2:75462279 - 75785697 on Build GRCh38 | Gain+Loss |
|
| dgv152e55 | Chr.2:75594376 - 75686819 on Build GRCh38 | Gain |
|
| nsv514988 | Chr.2:75632582 - 75690230 on Build GRCh38 | Loss |
|
| dgv5n43 | Chr.2:75591363 - 75680225 on Build GRCh38 | Gain |
|
| nsv441762 | Chr.2:75594779 - 75687246 on Build GRCh38 | Gain |
|
| nsv819219 | Chr.2:75662623 - 75689964 on Build GRCh38 | Gain |
|
| nsv834263 | Chr.2:75617574 - 75780540 on Build GRCh38 | Gain |
|
| esv3303063 | Chr.2:75593915 - 75690043 on Build GRCh38 | Tandem Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200111583] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map