Assay Details
Target Gene Details
Entrez Gene ID: | 400961 |
Gene Name: | poly(A) binding protein interacting protein 2B |
Gene Aliases: |
- |
Location: |
Chr.2:71182738-71227110 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAIP2B | NM_020459.1 | 4 | 1848 | NP_065192.1 |
| XM_005264310.3 | 5 | 1778 | XP_005264367.1 | |
| XM_005264311.4 | 4 | 1824 | XP_005264368.1 | |
| XM_011532842.2 | 4 | 1818 | XP_011531144.1 | |
| XM_011532843.2 | 4 | 1820 | XP_011531145.1 | |
| AB032981.1 | 4 | 1828 | ||
| AL110211.1 | 1 | 857 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834256 | Chr.2:71029484 - 71202682 on Build GRCh38 | Gain |
|
| nsv508148 | Chr.2:71120178 - 71201982 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map