Assay Details
Target Gene Details
Entrez Gene ID: | 843 |
Gene Name: | caspase 10 |
Gene Aliases: |
ALPS2, FLICE2, MCH4 |
Location: |
Chr.2:201182881-201229406 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP10 | NM_001206524.1 | NP_001193453.1 | ||
| NM_001206542.1 | NP_001193471.1 | |||
| NM_001230.4 | NP_001221.2 | |||
| NM_032974.4 | NP_116756.2 | |||
| NM_032976.3 | NP_116758.1 | |||
| NM_032977.3 | NP_116759.2 | |||
| XM_005246907.2 | XP_005246964.1 | |||
| XM_006712796.3 | XP_006712859.1 | |||
| XM_011511990.2 | XP_011510292.1 | |||
| AF111344.1 | AAD28402.1 | |||
| AF111345.1 | AAD28403.1 | |||
| AJ487678.1 | CAD32371.1 | |||
| AJ487679.1 | CAD32372.1 | |||
| AK304859.1 | ||||
| BC042844.1 | AAH42844.1 | |||
| KU177959.1 | ||||
| KU177960.1 | ||||
| U60519.1 | AAC50644.1 | |||
| U86214.1 | AAB46730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160898 | Chr.2:201196327 - 201213771 on Build GRCh38 | Deletion |
|
| nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |
|
| esv3593974 | Chr.2:201198802 - 201212212 on Build GRCh38 | Loss |
|
| nsv821874 | Chr.2:201200600 - 201210595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map