Assay Details
Target Gene Details
Entrez Gene ID: | 11037 |
Gene Name: | stonin 1 |
Gene Aliases: |
SALF, SBLF, STN1, STNB1 |
Location: |
Chr.2:48530169-48598515 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STON1 | NM_001198595.1 | NP_001185524.1 | ||
| NM_006873.3 | NP_006864.2 | |||
| AK314464.1 | ||||
| CR936807.1 | ||||
| DB288942.1 |
Target Gene Details
Entrez Gene ID: | 286749 |
Gene Name: | STON1-GTF2A1L readthrough |
Gene Aliases: |
SALF |
Location: |
Chr.2:48529925-48776517 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STON1-GTF2A1L | NM_001198593.1 | NP_001185522.1 | ||
| AK309740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv990475 | Chr.2:48554390 - 48558015 on Build GRCh38 | Deletion |
|
| esv2354047 | Chr.2:48554511 - 48557934 on Build GRCh38 | Deletion |
|
| esv8396 | Chr.2:48554545 - 48557934 on Build GRCh38 | Loss |
|
| nsv833981 | Chr.2:48478305 - 48651040 on Build GRCh38 | Loss |
|
| nsv1002793 | Chr.2:48544081 - 49239411 on Build GRCh38 | Gain |
|
| esv3556321 | Chr.2:48554573 - 48557822 on Build GRCh38 | Deletion |
|
| esv2631205 | Chr.2:48554103 - 48558672 on Build GRCh38 | Deletion |
|
| nsv1000419 | Chr.2:48539818 - 48568188 on Build GRCh38 | Loss |
|
| nsv581776 | Chr.2:48536243 - 48685880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map