Assay Details
Target Gene Details
Entrez Gene ID: | 102724965 |
Gene Name: | collagen alpha-2(I) chain-like |
Gene Aliases: |
- |
Location: |
Chr.2:45648733-45652047 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724965 | XM_011533229.2 | 3 | 1239 | XP_011531531.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3804n100 | Chr.2:45180603 - 45750333 on Build GRCh38 | Gain |
|
| esv2751894 | Chr.2:45141551 - 45772680 on Build GRCh38 | Gain |
|
| dgv1119e212 | Chr.2:45181796 - 45749281 on Build GRCh38 | Gain |
|
| nsv1013787 | Chr.2:45541783 - 45738705 on Build GRCh38 | Gain |
|
| esv2762545 | Chr.2:45419685 - 46137453 on Build GRCh38 | Gain |
|
| nsv581741 | Chr.2:45622938 - 45681498 on Build GRCh38 | Gain |
|

Genomic Map