Assay Details
Target Gene Details
Entrez Gene ID: | 129563 |
Gene Name: | DIS3 like 3'-5' exoribonuclease 2 |
Gene Aliases: |
FAM6A, PRLMNS, hDIS3L2 |
Location: |
Chr.2:231961583-232343968 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIS3L2 | NM_001257281.1 | NP_001244210.1 | ||
| NM_001257282.1 | NP_001244211.1 | |||
| NM_152383.4 | NP_689596.4 | |||
| NR_046476.1 | ||||
| NR_046477.1 | ||||
| AF443854.1 | AAP97321.1 | |||
| AK094293.1 | BAC04324.1 | |||
| AK295363.1 | ||||
| BC026166.1 | AAH26166.1 | |||
| BC036113.1 | AAH36113.2 | |||
| BX648325.1 | CAH10545.1 | |||
| DA058505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428408 | Chr.2:231672114 - 231968956 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map