Assay Details
Target Gene Details
Entrez Gene ID: | 100507098 |
Gene Name: | ADAMTS9 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:64684870-65011468 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS9-AS2 | NR_038264.1 | |||
| AK057923.1 | ||||
| AL832788.1 | ||||
| DA927615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834718 | Chr.3:64564777 - 64758878 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114707446] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map