Assay Details
Target Gene Details
Entrez Gene ID: | 8893 |
Gene Name: | eukaryotic translation initiation factor 2B subunit epsilon |
Gene Aliases: |
CACH, CLE, EIF-2B, EIF2Bepsilon, LVWM |
Location: |
Chr.3:184134435-184145311 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B5 | NM_003907.2 | NP_003898.2 | ||
| XM_011513265.1 | XP_011511567.1 | |||
| XM_011513266.2 | XP_011511568.1 | |||
| AB208821.1 | BAD92058.1 | |||
| AK091646.1 | BAC03712.1 | |||
| AK310207.1 | ||||
| AL832179.1 | ||||
| BC013590.1 | AAH13590.1 | |||
| U23028.1 | AAC50646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map