Assay Details
Target Gene Details
Entrez Gene ID: | 100287362 |
Gene Name: | protease, serine 46 |
Gene Aliases: |
TESSP6 |
Location: |
Chr.3:46719583-46736431 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS46 | NM_001205271.1 | NP_001192200.1 | ||
| AW183721.1 | ||||
| DB058077.1 | ||||
| DB099248.1 | ||||
| DY654973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516623 | Chr.3:46638573 - 46811189 on Build GRCh38 | Loss |
|
| nsv590179 | Chr.3:46687169 - 46748907 on Build GRCh38 | Loss |
|
| dgv4730n100 | Chr.3:46729944 - 46808087 on Build GRCh38 | Loss |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map