Assay Details
Target Gene Details
Entrez Gene ID: | 100505641 |
Gene Name: | FGD5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:14942779-14948441 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD5-AS1 | NR_046251.1 | 2 | 3339 | |
| NR_046252.1 | 2 | 3329 | ||
| NR_046253.1 | 2 | 3306 | ||
| NR_046254.1 | 2 | 3302 | ||
| NR_046255.1 | 2 | 3065 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759130 | Chr.3:14869763 - 15121196 on Build GRCh38 | Gain |
|
| nsv834620 | Chr.3:14912162 - 15093316 on Build GRCh38 | Gain |
|
| esv3595377 | Chr.3:14912760 - 14994553 on Build GRCh38 | Gain |
|

Genomic Map