Assay Details
Target Gene Details
Entrez Gene ID: | 51304 |
Gene Name: | zinc finger DHHC-type containing 3 |
Gene Aliases: |
DHHC-3, GODZ, ZNF373 |
Location: |
Chr.3:44915261-44976190 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC3 | NM_001135179.1 | 7 | 11629 | NP_001128651.1 |
| NM_016598.2 | 8 | 11713 | NP_057682.1 | |
| XM_005265209.1 | 8 | 3009 | XP_005265266.1 | |
| XM_005265212.2 | 7 | 2933 | XP_005265269.1 | |
| XM_006713189.1 | 8 | 3027 | XP_006713252.1 | |
| XM_006713194.3 | 7 | 2908 | XP_006713257.1 | |
| XM_011533797.1 | 9 | 3111 | XP_011532099.1 | |
| XM_011533800.2 | 9 | 3104 | XP_011532102.1 | |
| XM_017006557.1 | 10 | 3280 | XP_016862046.1 | |
| XM_017006558.1 | 11 | 3326 | XP_016862047.1 | |
| XM_017006559.1 | 10 | 3375 | XP_016862048.1 | |
| XM_017006560.1 | 9 | 3196 | XP_016862049.1 | |
| XM_017006561.1 | 9 | 3178 | XP_016862050.1 | |
| XM_017006562.1 | 8 | 3094 | XP_016862051.1 | |
| XM_017006565.1 | 10 | 3261 | XP_016862054.1 | |
| XM_017006568.1 | 8 | 2687 | XP_016862057.1 | |
| XM_017006570.1 | 8 | 3081 | XP_016862059.1 | |
| AK000216.1 | 1 | 765 | ||
| AK127837.1 | 1 | 3187 | ||
| AL832057.1 | 1 | 2148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834675 | Chr.3:44805220 - 44966381 on Build GRCh38 | Gain |
|
| esv3596040 | Chr.3:44911009 - 44916266 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map