Assay Details
Target Gene Details
Entrez Gene ID: | 131669 |
Gene Name: | urocanate hydratase 1 |
Gene Aliases: |
HMFN0320, UROCD |
Location: |
Chr.3:126481165-126517773 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UROC1 | NM_001165974.1 | NP_001159446.1 | ||
| NM_144639.2 | NP_653240.1 | |||
| AB075869.1 | BAD38651.1 | |||
| AK055862.1 | BAB71032.1 | |||
| BC115405.1 | ||||
| BC115406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829717 | Chr.3:126397190 - 126583201 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116675558] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map