Assay Details
Target Gene Details
Entrez Gene ID: | 54899 |
Gene Name: | PX domain containing serine/threonine kinase like |
Gene Aliases: |
MONaKA |
Location: |
Chr.3:58332868-58426127 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528147 | Chr.3:57929747 - 58384450 on Build GRCh38 | Gain |
|
| nsv1001814 | Chr.3:58158881 - 58354197 on Build GRCh38 | Gain |
|
| dgv4748n100 | Chr.3:58316064 - 58399467 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map