Assay Details
Target Gene Details
Entrez Gene ID: | 54965 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class X |
Gene Aliases: |
PIG-X |
Location: |
Chr.3:196712374-196736007 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGX | NM_001166304.1 | NP_001159776.1 | ||
| NM_017861.3 | NP_060331.3 | |||
| AA879458.1 | ||||
| AK000529.1 | BAA91233.1 | |||
| AK294340.1 | 1 | 276 | ||
| BC022542.1 | AAH22542.1 | |||
| BG715890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014526 | Chr.3:196645551 - 196733298 on Build GRCh38 | Gain |
|
| nsv829828 | Chr.3:196541250 - 196736770 on Build GRCh38 | Loss |
|
| nsv1009975 | Chr.3:196680410 - 196822871 on Build GRCh38 | Gain |
|
| nsv1001045 | Chr.3:196703082 - 196900384 on Build GRCh38 | Gain |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| nsv1014692 | Chr.3:196515378 - 196804953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map