Assay Details
Target Gene Details
Entrez Gene ID: | 57406 |
Gene Name: | abhydrolase domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.3:58237502-58294736 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD6 | NM_001320126.1 | NP_001307055.1 | ||
| NM_020676.6 | NP_065727.4 | |||
| XM_005265334.3 | XP_005265391.1 | |||
| XM_005265335.3 | XP_005265392.1 | |||
| AF225418.1 | AAG09720.1 | |||
| AK122983.1 | ||||
| AK172797.1 | BAD18771.1 | |||
| AK225942.1 | ||||
| AK313168.1 | ||||
| BC001698.2 | AAH01698.1 | |||
| DA266414.1 | ||||
| DB171182.1 | ||||
| HY038261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834703 | Chr.3:58072791 - 58298770 on Build GRCh38 | Gain |
|
| nsv528147 | Chr.3:57929747 - 58384450 on Build GRCh38 | Gain |
|
| nsv1001814 | Chr.3:58158881 - 58354197 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114623404,rs74804144] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map