Assay Details
Target Gene Details
Entrez Gene ID: | 100874330 |
Gene Name: | long intergenic non-protein coding RNA 1014 |
Gene Aliases: |
KCNMB2-IT1 |
Location: |
Chr.3:178419201-178457305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01014 | NR_046786.1 | |||
| BX105665.1 |

Genomic Map