Assay Details
Target Gene Details
Entrez Gene ID: | 3568 |
Gene Name: | interleukin 5 receptor subunit alpha |
Gene Aliases: |
CD125, CDw125, HSIL5R3, IL5R |
Location: |
Chr.3:3066324-3110414 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL5RA | NM_000564.4 | 13 | 4314 | NP_000555.2 |
| NM_001243099.1 | 11 | 4160 | NP_001230028.1 | |
| NM_175726.3 | 12 | 4245 | NP_783853.1 | |
| XM_011533677.2 | 13 | 4302 | XP_011531979.1 | |
| XM_011533678.2 | 13 | 4287 | XP_011531980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
| esv3893627 | Chr.3:3063890 - 3069774 on Build GRCh38 | Loss |
|
| nsv1013843 | Chr.3:3058965 - 3088961 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map