Assay Details
Target Gene Details
Entrez Gene ID: | 2932 |
Gene Name: | glycogen synthase kinase 3 beta |
Gene Aliases: |
- |
Location: |
Chr.3:119821321-120094417 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSK3B | NM_001146156.1 | 11 | 5933 | NP_001139628.1 |
| NM_002093.3 | 12 | 5972 | NP_002084.2 | |
| XM_006713610.2 | 11 | 5359 | XP_006713673.1 | |
| XM_006713611.2 | 10 | 5320 | XP_006713674.1 | |
| BX640779.1 | 1 | 2683 |
More Information
Set Membership: |
|

Genomic Map