Assay Details
Target Gene Details
Entrez Gene ID: | 100505641 |
Gene Name: | FGD5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:14942779-14948441 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD5-AS1 | NR_046251.1 | 1 | 148 | |
| AK096279.1 | 1 | 131 | ||
| DA993974.1 | 1 | 148 |
Target Gene Details
Entrez Gene ID: | 7182 |
Gene Name: | nuclear receptor subfamily 2 group C member 2 |
Gene Aliases: |
TAK1, TR4 |
Location: |
Chr.3:14947584-15049279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2C2 | NM_001291694.1 | NP_001278623.1 | ||
| NM_003298.4 | NP_003289.2 | |||
| XM_011534058.2 | XP_011532360.1 | |||
| XM_011534061.2 | XP_011532363.1 | |||
| XM_011534063.2 | XP_011532365.1 | |||
| XM_011534064.2 | XP_011532366.1 | |||
| XM_011534065.2 | XP_011532367.1 | |||
| AK290590.1 | ||||
| L27586.1 | AAA21474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759130 | Chr.3:14869763 - 15121196 on Build GRCh38 | Gain |
|
| nsv834620 | Chr.3:14912162 - 15093316 on Build GRCh38 | Gain |
|
| esv3595377 | Chr.3:14912760 - 14994553 on Build GRCh38 | Gain |
|
| nsv1074573 | Chr.3:14947792 - 14948793 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map