Assay Details
Target Gene Details
Entrez Gene ID: | 2917 |
Gene Name: | glutamate metabotropic receptor 7 |
Gene Aliases: |
GLUR7, GPRC1G, MGLU7, MGLUR7, PPP1R87 |
Location: |
Chr.3:6861097-7743032 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM7 | NM_000844.3 | NP_000835.1 | ||
| NM_181874.2 | NP_870989.1 | |||
| XM_017006272.1 | XP_016861761.1 | |||
| XM_017006273.1 | XP_016861762.1 | |||
| AB209214.1 | BAD92451.1 | |||
| AK097817.1 | ||||
| AK312488.1 | ||||
| BC136458.1 | ||||
| BC136459.1 | ||||
| X94552.1 | CAA64245.1 |
Target Gene Details
Entrez Gene ID: | 100873937 |
Gene Name: | GRM7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:7519741-7535284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM7-AS1 | NR_046606.1 | |||
| AI222038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595196 | Chr.3:7507741 - 7629043 on Build GRCh38 | Gain |
|
| nsv1006252 | Chr.3:6707783 - 7544588 on Build GRCh38 | Gain |
|
| esv3595197 | Chr.3:7509066 - 7752803 on Build GRCh38 | Gain |
|
| esv3595198 | Chr.3:7518416 - 7521160 on Build GRCh38 | Loss |
|
| nsv589568 | Chr.3:6713211 - 7549718 on Build GRCh38 | Gain |
|
| esv2759128 | Chr.3:7482865 - 7522616 on Build GRCh38 | Gain |
|
| nsv834610 | Chr.3:7457127 - 7668584 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map