Assay Details
Target Gene Details
Entrez Gene ID: | 22907 |
Gene Name: | DEAH-box helicase 30 |
Gene Aliases: |
DDX30, RETCOR |
Location: |
Chr.3:47802909-47850196 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834679 | Chr.3:47707985 - 47882516 on Build GRCh38 | Loss |
|
| nsv508916 | Chr.3:47753956 - 47883831 on Build GRCh38 | Insertion |
|
| nsv3808 | Chr.3:47762613 - 47805200 on Build GRCh38 | Insertion |
|
| nsv231 | Chr.3:47775571 - 47804097 on Build GRCh38 | Insertion |
|
| nsv10270 | Chr.3:47781116 - 47804088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map