Assay Details
Target Gene Details
Entrez Gene ID: | 100874330 |
Gene Name: | long intergenic non-protein coding RNA 1014 |
Gene Aliases: |
KCNMB2-IT1 |
Location: |
Chr.3:178419201-178457305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01014 | NR_046786.1 | |||
| BX105665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525869 | Chr.3:178401581 - 178436727 on Build GRCh38 | Gain |
|
| nsv513734 | Chr.3:178398576 - 178439042 on Build GRCh38 | Insertion |
|
| nsv4136 | Chr.3:178386237 - 178431491 on Build GRCh38 | Deletion |
|
| nsv4137 | Chr.3:178420749 - 178437917 on Build GRCh38 | Insertion |
|

Genomic Map