Assay Details
Target Gene Details
Entrez Gene ID: | 51095 |
Gene Name: | tRNA nucleotidyl transferase 1 |
Gene Aliases: |
CCA1, CGI-47, MtCCA, RPEM, SIFD |
Location: |
Chr.3:3126916-3153435 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRNT1 | NM_001302946.1 | NP_001289875.1 | ||
| NM_182916.2 | NP_886552.2 | |||
| XM_011533776.2 | XP_011532078.1 | |||
| XM_011533777.2 | XP_011532079.1 | |||
| XM_011533778.2 | XP_011532080.1 | |||
| AB063105.1 | BAB70662.1 | |||
| AF151805.1 | AAD34042.1 | |||
| AK290411.1 | ||||
| AL834397.1 | CAD39059.1 | |||
| DB032432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv787n67 | Chr.3:3132564 - 3135456 on Build GRCh38 | Loss |
|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |
|
| esv3595052 | Chr.3:3131503 - 3135391 on Build GRCh38 | Loss |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map