Assay Details
Target Gene Details
Entrez Gene ID: | 80852 |
Gene Name: | glutamate receptor interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.3:14489111-14602996 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIP2 | NM_001080423.3 | 24 | 3232 | NP_001073892.3 |
| XM_011534141.2 | 24 | 3314 | XP_011532443.1 | |
| XM_011534142.2 | 24 | 3226 | XP_011532444.1 | |
| XM_011534143.2 | 23 | 3188 | XP_011532445.1 | |
| XM_011534145.2 | 18 | 2960 | XP_011532447.1 | |
| XM_011534146.2 | 23 | 3793 | XP_011532448.1 | |
| AB051506.2 | 24 | 3218 | ||
| AK024507.1 | 11 | 1854 | BAB15797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion |
|
| nsv518216 | Chr.3:14490517 - 14502161 on Build GRCh38 | Loss |
|
| nsv3712 | Chr.3:14467853 - 14501724 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map