Assay Details
Target Gene Details
Entrez Gene ID: | 51185 |
Gene Name: | cereblon |
Gene Aliases: |
MRT2, MRT2A |
Location: |
Chr.3:3148490-3179717 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRBN | NM_001173482.1 | NP_001166953.1 | ||
| NM_016302.3 | NP_057386.2 | |||
| XM_005265202.3 | XP_005265259.1 | |||
| XM_011533791.2 | XP_011532093.1 | |||
| XM_011533793.2 | XP_011532095.1 | |||
| XM_011533794.2 | XP_011532096.1 | |||
| AF117230.1 | AAF17211.1 | |||
| AK027507.1 | BAB55162.1 | |||
| AK058191.1 | ||||
| AK301423.1 | ||||
| AK312577.1 | ||||
| BC017419.1 | AAH17419.1 | |||
| BC067811.1 | AAH67811.1 | |||
| CR627060.1 | CAH10361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
| nsv834604 | Chr.3:3147083 - 3361983 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77784443] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map