Assay Details
Target Gene Details
Entrez Gene ID: | 100130872 |
Gene Name: | uncharacterized LOC100130872 |
Gene Aliases: |
- |
Location: |
Chr.4:1195783-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130872 | NR_024569.1 | |||
| AK091779.1 | 1 | 740 | BAC03745.1 | |
| CR627052.1 |
Target Gene Details
Entrez Gene ID: | 101928548 |
Gene Name: | uncharacterized LOC101928548 |
Gene Aliases: |
- |
Location: |
Chr.4:1200379-1201262 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928548 | XM_005272308.3 | 1 | 586 | XP_005272365.1 |
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPON2 | NM_001199021.1 | NP_001185950.1 | ||
| CB104725.1 | ||||
| DA201089.1 | ||||
| DB249869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss |
|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| dgv636n27 | Chr.4:1165892 - 1261142 on Build GRCh38 | Loss |
|
| nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion |
|
| nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion |
|
| nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map