Assay Details
Target Gene Details
Entrez Gene ID: | 2982 |
Gene Name: | guanylate cyclase 1 soluble subunit alpha |
Gene Aliases: |
GC-SA3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, MYMY6 |
Location: |
Chr.4:155666710-155737062 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602766 | Chr.4:155675673 - 155690759 on Build GRCh38 | Loss |
|
| nsv437445 | Chr.4:155663245 - 155693802 on Build GRCh38 | Loss |
|
| esv2759290 | Chr.4:155685754 - 155689688 on Build GRCh38 | Loss |
|
| nsv519870 | Chr.4:155676929 - 155690151 on Build GRCh38 | Loss |
|
| esv3894054 | Chr.4:155675907 - 155689688 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79715470] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map