Assay Details
Target Gene Details
Entrez Gene ID: | 389207 |
Gene Name: | glutaredoxin and cysteine rich domain containing 1 |
Gene Aliases: |
DFNB25, PPP1R88 |
Location: |
Chr.4:42893266-43030658 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRXCR1 | NM_001080476.2 | 1 | 83 | NP_001073945.1 |
More Information
Set Membership: |
|

Genomic Map