Assay Details
Target Gene Details
Entrez Gene ID: | 389207 |
Gene Name: | glutaredoxin and cysteine rich domain containing 1 |
Gene Aliases: |
DFNB25, PPP1R88 |
Location: |
Chr.4:42893266-43030658 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRXCR1 | NM_001080476.2 | NP_001073945.1 | ||
| DT932651.1 | ||||
| DY654338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv872n67 | Chr.4:42903094 - 42903841 on Build GRCh38 | Loss |
|
| esv2727494 | Chr.4:42903128 - 42903831 on Build GRCh38 | Deletion |
|
| esv3563781 | Chr.4:42903092 - 42903896 on Build GRCh38 | Deletion |
|
| esv7471 | Chr.4:42902957 - 42904056 on Build GRCh38 | Loss |
|
| esv5262 | Chr.4:42903115 - 42903916 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143492369] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map