Assay Details
Target Gene Details
Entrez Gene ID: | 152992 |
Gene Name: | tRNA methyltransferase 44 homolog (S. cerevisiae) |
Gene Aliases: |
C4orf23, METTL19, TRM44 |
Location: |
Chr.4:8440774-8512531 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT44 | NM_152544.2 | NP_689757.2 | ||
| XM_005247950.3 | XP_005248007.1 | |||
| XM_011513406.2 | XP_011511708.1 | |||
| XM_011513407.2 | XP_011511709.1 | |||
| XM_011513408.1 | XP_011511710.1 | |||
| XM_011513409.2 | XP_011511711.1 | |||
| XM_011513410.2 | XP_011511712.1 | |||
| XM_017007811.1 | XP_016863300.1 | |||
| BC035655.1 | AAH35655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593638 | Chr.4:8371217 - 8456667 on Build GRCh38 | Loss |
|
| nsv1002794 | Chr.4:8108356 - 8489207 on Build GRCh38 | Gain |
|
| nsv1005698 | Chr.4:8434559 - 10248775 on Build GRCh38 | Gain |
|
| nsv829854 | Chr.4:8401194 - 8504680 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map