Assay Details
Target Gene Details
Entrez Gene ID: | 150159 |
Gene Name: | solute carrier family 9 member B1 |
Gene Aliases: |
NHA1, NHEDC1 |
Location: |
Chr.4:102885048-103019739 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9B1 | NM_001100874.2 | NP_001094344.1 | ||
| NM_139173.3 | NP_631912.2 | |||
| NR_047513.1 | ||||
| NR_047515.1 | ||||
| XM_006714093.3 | XP_006714156.1 | |||
| XM_011531622.1 | XP_011529924.1 | |||
| XM_011531623.1 | XP_011529925.1 | |||
| AF447585.1 | AAM22868.1 | |||
| AK313592.1 | ||||
| AY461581.1 | ||||
| BC110794.1 | AAI10795.1 | |||
| BC132712.1 | ||||
| BC136966.1 | ||||
| DQ003059.1 | AAY21808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461598 | Chr.4:102886563 - 103032232 on Build GRCh38 | Loss |
|
| esv3569636 | Chr.4:102970540 - 103009732 on Build GRCh38 | Loss |
|
| esv2759270 | Chr.4:102859548 - 103136164 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map