Assay Details
Target Gene Details
Entrez Gene ID: | 132430 |
Gene Name: | poly(A) binding protein cytoplasmic 4 like |
Gene Aliases: |
- |
Location: |
Chr.4:133948459-134201748 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PABPC4L |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602301 | Chr.4:134007930 - 134076602 on Build GRCh38 | Loss |
|
| dgv9274n54 | Chr.4:133948569 - 134063406 on Build GRCh38 | Loss |
|
| esv3569696 | Chr.4:133997798 - 134270024 on Build GRCh38 | Loss |
|
| nsv1023249 | Chr.4:133946159 - 134064530 on Build GRCh38 | Loss |
|
| dgv9275n54 | Chr.4:133985106 - 134274527 on Build GRCh38 | Loss |
|
| esv3602299 | Chr.4:133999957 - 134149092 on Build GRCh38 | Loss |
|
| dgv5396n100 | Chr.4:133889406 - 134023164 on Build GRCh38 | Loss |
|
| dgv5397n100 | Chr.4:133997524 - 134267621 on Build GRCh38 | Loss |
|
| nsv520680 | Chr.4:134001210 - 134274527 on Build GRCh38 | Loss |
|
| nsv1015565 | Chr.4:133995284 - 134147172 on Build GRCh38 | Loss |
|

Genomic Map