Assay Details
Target Gene Details
Entrez Gene ID: | 2982 |
Gene Name: | guanylate cyclase 1 soluble subunit alpha |
Gene Aliases: |
GC-SA3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, MYMY6 |
Location: |
Chr.4:155666710-155737062 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602766 | Chr.4:155675673 - 155690759 on Build GRCh38 | Loss |
|
| nsv437445 | Chr.4:155663245 - 155693802 on Build GRCh38 | Loss |
|
| nsv442925 | Chr.4:155673116 - 155676843 on Build GRCh38 | Loss |
|
| nsv514260 | Chr.4:155673238 - 155676646 on Build GRCh38 | Loss |
|
| esv2760878 | Chr.4:155640759 - 155680884 on Build GRCh38 | Loss |
|
| nsv1022201 | Chr.4:155657250 - 155676831 on Build GRCh38 | Loss |
|
| esv2674234 | Chr.4:155673048 - 155677151 on Build GRCh38 | Deletion |
|
| esv3602765 | Chr.4:155673010 - 155677231 on Build GRCh38 | Loss |
|
| esv23465 | Chr.4:155673053 - 155677130 on Build GRCh38 | Loss |
|
| esv3894054 | Chr.4:155675907 - 155689688 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map