Assay Details
Target Gene Details
Entrez Gene ID: | 4043 |
Gene Name: | LDL receptor related protein associated protein 1 |
Gene Aliases: |
A2MRAP, A2RAP, HBP44, MRAP, MYP23, RAP, alpha-2-MRAP |
Location: |
Chr.4:3503597-3532497 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRPAP1 | NM_002337.3 | NP_002328.1 | ||
| NR_110005.1 | ||||
| AK222504.1 | BAD96224.1 | |||
| AK222527.1 | BAD96247.1 | |||
| AK292321.1 | ||||
| AK311600.1 | ||||
| AK312698.1 | ||||
| BC105074.1 | AAI05075.1 | |||
| BC112067.1 | AAI12068.1 | |||
| BX647984.1 | ||||
| M63959.1 | AAA51553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998670 | Chr.4:3452060 - 3907808 on Build GRCh38 | Gain |
|
| nsv829841 | Chr.4:3500691 - 3661692 on Build GRCh38 | Loss |
|
| nsv428435 | Chr.4:3385046 - 3621594 on Build GRCh38 | Gain |
|
| nsv949767 | Chr.4:3509374 - 3545773 on Build GRCh38 | Deletion |
|
| esv2759219 | Chr.4:3392617 - 4349259 on Build GRCh38 | Gain |
|
| nsv508992 | Chr.4:3455794 - 3533692 on Build GRCh38 | Insertion |
|
| nsv10426 | Chr.4:3477763 - 3565983 on Build GRCh38 | Gain+Loss |
|
| nsv829840 | Chr.4:3442489 - 3621304 on Build GRCh38 | Loss |
|
| nsv470004 | Chr.4:3428129 - 3582646 on Build GRCh38 | Loss |
|
| nsv461171 | Chr.4:3484059 - 3520956 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73793979] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map