Assay Details
Target Gene Details
Entrez Gene ID: | 550113 |
Gene Name: | uncharacterized LOC550113 |
Gene Aliases: |
- |
Location: |
Chr.4:68053508-68073233 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC550113 | NR_046116.1 | |||
| AI001926.1 |
Target Gene Details
Entrez Gene ID: | 389208 |
Gene Name: | transmembrane protease, serine 11F |
Gene Aliases: |
- |
Location: |
Chr.4:68053198-68129869 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS11F | NM_207407.2 | NP_997290.2 | ||
| XM_011531935.1 | XP_011530237.1 | |||
| AK122625.1 | BAC85495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759257 | Chr.4:67911288 - 68163195 on Build GRCh38 | Gain |
|
| nsv818242 | Chr.4:67945419 - 68132435 on Build GRCh38 | Gain |
|
| nsv515010 | Chr.4:68053175 - 68129695 on Build GRCh38 | Loss |
|
| esv22957 | Chr.4:67923012 - 68150383 on Build GRCh38 | Gain |
|
| nsv10514 | Chr.4:67923797 - 68150168 on Build GRCh38 | Gain |
|
| nsv469718 | Chr.4:67952598 - 68109856 on Build GRCh38 | Gain |
|
| esv3600857 | Chr.4:67925571 - 68142494 on Build GRCh38 | Gain |
|
| nsv1007376 | Chr.4:67827765 - 68142484 on Build GRCh38 | Gain |
|
| esv35010 | Chr.4:67925116 - 68152450 on Build GRCh38 | Gain |
|
| nsv433367 | Chr.4:67931662 - 68147283 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80059776] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map