Assay Details
Target Gene Details
Entrez Gene ID: | 285527 |
Gene Name: | FRY like transcription coactivator |
Gene Aliases: |
AF4p12, KIAA0826, MOR2 |
Location: |
Chr.4:48497363-48780312 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 57 - Exon 57 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRYL | NM_015030.1 | NP_055845.1 | ||
| XM_005248090.4 | XP_005248147.1 | |||
| XM_005248093.4 | XP_005248150.1 | |||
| XM_011513680.2 | XP_011511982.1 | |||
| XM_011513681.2 | XP_011511983.1 | |||
| XM_011513682.2 | XP_011511984.1 | |||
| XM_011513683.2 | XP_011511985.1 | |||
| XM_011513684.2 | XP_011511986.1 | |||
| XM_017008046.1 | XP_016863535.1 | |||
| AB020633.1 | BAA74849.1 | |||
| AK127307.1 | ||||
| AL833170.1 | ||||
| U80082.1 | AAD00351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829925 | Chr.4:48465676 - 48637325 on Build GRCh38 | Gain+Loss |
|
| nsv470034 | Chr.4:48425756 - 48603611 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map